missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, Eluent additive for LC-MS, Honeywell Fluka™

Product Code. 15658430
Click to view available options
Quantity:
50 mL
10 x 1 mL
1 L
Packaging:
Ampoule
Glass Bottle
Plastic Bottle

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Melting Point 8.2°C to 8.4°C (literature)
Boiling Point 100.4°C
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Linear Formula H2O
UN Number UN1779
Beilstein 1209246
Formula Weight 46.03
Percent Purity >97.5%
Grade LC-MS
Density 1.22 g/mL at 25 °C(lit.)
Quantity 10 x 1 mL
Explosion Limit 0.57
Flash Point 49.5°C
Ignition Point 1004°F
Packaging Ampoule
Vapor Density 1.6 (vs air)
Vapor Pressure 44.8 mmHg (20°C)
Chemical Name or Material Formic acid, Eluent additive for LC-MS
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.