missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Forskolin, (from Coleus forskohlii), MP Biomedicals™
36.05 € - 585.00 €
Chemical Identifiers
CAS | 66575-29-9 |
---|---|
Molecular Formula | C22H34O7 |
Molecular Weight (g/mol) | 410.507 |
MDL Number | MFCD00082317 |
InChI Key | OHCQJHSOBUTRHG-KGGHGJDLSA-N |
Synonym | forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish |
PubChem CID | 47936 |
ChEBI | CHEBI:42471 |
IUPAC Name | [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate |
SMILES | CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11446071
|
MP Biomedicals
0219066901 |
1 mg |
36.05 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11456071
|
MP Biomedicals
0219066905 |
5 mg |
163.00 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11466071
|
MP Biomedicals
0219066910 |
10 mg |
258.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11476071
|
MP Biomedicals
0219066925 |
25 mg |
585.00 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
66575-29-9 | |
410.507 | |
OHCQJHSOBUTRHG-KGGHGJDLSA-N | |
47936 | |
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate |
C22H34O7 | |
MFCD00082317 | |
forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish | |
CHEBI:42471 | |
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O |
Specifications
66575-29-9 | |
0.9775 | |
MFCD00082317 | |
OHCQJHSOBUTRHG-KGGHGJDLSA-N | |
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate | |
47936 | |
410.5 | |
Forskolin, from Coleus forskohlii |
White | |
C22H34O7 | |
forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish | |
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O | |
410.507 | |
CHEBI:42471 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 266-410-9
Recommended Storage : Store at -0°C. Store Desiccated.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Forskoli
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title