Learn More
Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringe
Able to be autoclaved at 134°C. Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringes come with an interchangeable piston.
Specifications
Autoclavable Autoclavable
Certifications/Compliance CE EN 46001
Material Borosilicate Glass
Description Amber graduation
Product Type All Glass Luer-lock Syringe
View More Specs
Description
Description
  • Complies with CE EN 46001 standards
  • Constructed from Durobax™ glass
  • With permanent amber graduation
  • Acid and alkali resistant
Specifications
Specifications
Autoclavable
Borosilicate Glass
All Glass Luer-lock Syringe
Luer Lock
CE EN 46001
Amber graduation
1/Ea.
Yes