missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Four elements solution - Ca, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical MS 010 100
This item is not returnable.
View return policy
Specifications
Four Elements | |
100 mL | |
1000 mg/L | |
Ca, Mg, K, Na |
Bottle | |
Liquid | |
ICP/MS Calibration Multi-element Standard Solution | |
2% HNO3 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1088668 | Certificate of Analysis | 05/03/2025 | MS 010 100 |
1079673 | Certificate of Analysis | 04/03/2025 | MS 010 100 |
1071524 | Certificate of Analysis | 04/03/2025 | MS 010 100 |
1012739 | Certificate of Analysis | 04/03/2025 | MS 010 100 |
1044337 | Certificate of Analysis | 04/03/2025 | MS 010 100 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Four elements solution - Ca, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™ >
Spot an opportunity for improvement?Share a Content Correction