missing translation for 'onlineSavingsMsg'
Learn More

FPA 124, Tocris Bioscience™

Product Code. 30001445
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Akt/PKB inhibitor

Akt/PKB inhibitor (IC50 = 100 nM) that interacts with the PH and kinase domains. Inhibits cell proliferation in various cancer cell lines in vitro and decreases NF-κB activity and tumor load in vivo.

Chemical Identifiers

CAS 902779-59-3
Molecular Formula C11H9Cl2CuN3O2S
Molecular Weight (g/mol) 381.718
InChI Key HZRXQEVFXXQYFS-UHFFFAOYSA-L
Synonym Dichloro[(2Z)-2-[(4-oxo-4H-1-benzopyran-3-yl)methylene]hydrazinecarbothioamide copper complex
PubChem CID 102586612
IUPAC Name dichlorocopper;[(4-oxochromen-3-yl)methylideneamino]thiourea
SMILES C1=CC=C2C(=C1)C(=O)C(=CO2)C=NNC(=S)N.Cl[Cu]Cl

Specifications

Quantity 10 mg
Formula Weight 381.73
Percent Purity >98%
Chemical Name or Material FPA 124
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'