Learn More
Cytiva Frac30 Model Fraction Collector
Used as a round fraction collector. Cytiva Frac30 Model Fraction Collector allows you to collect up to 30 fractions.
Brand:  Cytiva 29023051
Product Code. 15279764
  • 1742.00 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Controlled via the AKTA protein purification system start touch screen instrument display
  • Also controlled by using UNICORN start software via computer
  • Collect up to 30 fractions
  • Supports four tube sizes, ranging from 1.5mL to 15mL
  • Fractions can be collected in volumes ranging from 0.5mL to 15mL
  • The entire fraction collector rack can be transferred as required
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fraction Collector