Learn More
Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules, BreakApart frame
Use Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules with Nunc™ C8 Breakable Modules.
84.20 € - 87.26 €
Specifications
Format Strip Plate
View More Specs
Products 3
Product Code Brand Product Type No. per Pack No. per Case Plate Compatibility Price Quantity & Availability  
Product Code Brand Product Type No. per Pack No. per Case Plate Compatibility Price Quantity & Availability  
10585211
View Documents
Thermo Scientific™
431615
Frame 5 60 Nunc™ C8 Breakable Modules
87.16 €
Pack of 60
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
10524641
View Documents
Thermo Scientific™
465404
Frame 5 60 Immuno Standard Modules
87.26 €
Pack of 60
Estimated Shipment: 21-03-2025
to see stock.
Add to basket
10733316
View Documents
Thermo Scientific™
9503110
Frame 50 50 Nunc™ C8 Breakable Modules This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Use Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules with Nunc™ C8 Breakable Modules. These frames include tools to facilitate removing the strips.
Warranty: 90 days
Specifications
Specifications
Strip Plate