missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ FreeStyle™ MAX Reagent
Animal origin-free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins
802.00 € - 87800.00 €
Specifications
Classification | Animal Origin-Free |
---|---|
Cell Type | Established Cell Lines |
Sample Type | Plasmid DNA |
Transfection Technique | Lipid-based Transfection |
High-throughput Compatibility | Not High-throughput Compatible (Manual) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10259172
|
Gibco™
16447100 |
1 mL |
802.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11503147
|
Gibco™
16447500 |
15 mL |
11220.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10489684
|
Gibco™
16447750 |
10 x 15 mL |
87800.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Allows highest expression levels and transfection rates with lowest cytotoxicity in bio-production applications
- Formulated for use with:
- FreeStyle 293-F Cells (suspension human embryonal kidney cells, Cat. No. R790-07) in serum-free FreeStyle 293 Expression Medium (Cat. No. 12338-018)
- FreeStyle CHO-S Cells (suspension Chinese Hamster Ovary cells, Cat. No. R800-07) in serum-free FreeStyle CHO Expression Medium (Cat. No. 12651-014)
- DG44 Cells (DHFR– suspension CHO cells) in CD DG44 Medium with 8 mM L-glutamine and 18 mL/L 10% Pluronic F-68 (Cat. No. A11000-01)
- FreeStyle™ MAX Reagent is a proprietary, animal origin–free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins
- Intended for use with the FreeStyle™ MAX 293 Expression System (Cat. No. K9000-10) and the FreeStyle™ MAX CHO Expression System (Cat. No. K9000-20).
Specifications
Animal Origin-Free | |
Plasmid DNA | |
Not High-throughput Compatible (Manual) | |
Transfection Reagent | |
Wet Ice |
Established Cell Lines | |
Lipid-based Transfection | |
Freestyle™ | |
Yes |
For Research Use Only. Not for use in diagnostic procedures.