missing translation for 'onlineSavingsMsg'
Learn More

Freeze Drying Ampules

Use these ampules for a variety of laboratory applications. Freeze Drying Ampules are manufactured neutral glass to the dimensions, tolerances and quality requirements of ISO 4142:2002, to meet all applications for re-usable glass tubes.

Brand:  Samco G225/02

 View more versions of this product

Product Code. 13131793

  • 559.00 € / Pack of 250
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use these ampules for a variety of laboratory applications. Freeze Drying Ampules are manufactured neutral glass to the dimensions, tolerances and quality requirements of ISO 4142:2002, to meet all applications for re-usable glass tubes.

  • Clear neutral glass vials for freeze drying
  • The top 11mm of the open ends are glazed with an internal neck diameter of 5.9mm to 6.1mm
  • Capacity and length are nominal
Specifications

Specifications

2.5 mL
Ampule
7 mm
Neutral Glass
Clear
154mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.