missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Storage Boxes

Store a variety of vials and tubes with these fiberboard storage boxes, for use in ultra-low temperature freezers.

Brand:  Thermo Scientific™ 820010

 View more versions of this product

Product Code. 13457238

  • 38.29 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Store a variety of vials and tubes with Thermo Scientific™ Storage Boxes, for use in ultra-low temperature freezers.

  • Sturdy, water-repellant boxes with covers for sample storage
  • Available in 2 and 3 in. high (5.1 and 7.6cm) with 5.25 × 5.25 in. (13.3 × 13.3cm) square bases
  • Box dividers included (excluding 5956)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
Square
Thermo Scientific Ultra-Low Temperature Freezers
Dividers
5.3 in.
5.3 in.
2 in.
1 kg
Polycarbonate
Cryo Storage Box
100 (1, 1.5, 2 mL Cryo Tubes)
5.3 x 5.3 x 2 in.
13.4 cm
13.4 cm
5.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.