Learn More
Brady™ FreezerBondz™ Polyester Autoclave and Cryogenic Labels for M710
FreezerBondz™ Label Printer Labels for use with Brady™ Printer M710.
Brand:  Brady™ 311982
 View more versions of this product
Product Code. 17837934
  • 89.20 € / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Good print smudge resistance, solvent resistance
  • High/low-temperature performance range of -196°C to +100°C
Specifications
Specifications
ASTM D 1000
Permanent Acrylic
White
Label
Single-side Printable
Cryogenic Label
General Identification, Laboratory Labeling
1 Label
500 Labels
-320°F to +230°F
0.0032 in.
12.7 x 25.4 mm
1 in.
25.4 mm
M7-17-492
None
Matte
Polyester
Thermal Transfer
Non-sterile
M710, BMP71
-196°C
Rectangle
-196°C to +110°C
0.081 mm
0.5 in.
25.4 mm