Learn More
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady™ handheld label maker.
Brand:  Brady™ 170897
 View more versions of this product
Product Code. 17891799
  • 95.25 € / Pack of 180
Explore more special offers
Description
Description
  • Ideal for laboratory use on curved surfaces including test tubes, vials and straw pipettes
  • Withstands chemicals, abrasions and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
Specifications
Specifications
RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Matte
Polyester
Labels
General Identification, Laboratory Labeling
180 Labels/Cartridge
0.0032 in.
0.25 in.
3.2 cm
M5-155-492
Black on White
Die-cut
Thermal Transfer
Non-sterile
BMP51, M511
-320°F to +230°F
0.25 x 1.25 in.
1.25 in.