Learn More
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady™ handheld label maker.
Brand:  Brady™ 170778
 View more versions of this product
Product Code. 17950730
  • 93.25 € / Pack of 240
Explore more special offers
Description
Description
  • Ideal for laboratory use on curved surfaces including test tubes, vials and straw pipettes
  • Withstands chemicals, abrasions and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
Specifications
Specifications
Not Autoclavable
M4-82-492
Black on White
Die-cut
Thermal Transfer
Non-sterile
BMP41, BMP51, M511
-320°F to +230°F
0.375 x 0.375 in.
1.9 in.
0.5 in.
RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Matte
Polyester
Labels
General Identification, Laboratory Labeling
240 Labels/Cartridge
0.0032 in.
0.375 in.
4.8 cm