Learn More
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady™ handheld label maker.
Brand:  Brady™ 170777
 View more versions of this product
Product Code. 17980690
  • 101.00 € / Each
Explore more special offers
Description
Description
  • Ideal for laboratory use on curved surfaces including test tubes, vials and straw pipettes
  • Withstands chemicals, abrasions and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
Specifications
Specifications
Not Autoclavable
M4C-500-492
Black on White
Continuous
Thermal Transfer
Non-sterile
BMP41, BMP51, M511
-320°F to +230°F
25 ft.
1.3 cm
RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Matte
Polyester
Labels
Component and Equipment Labeling, Data and Telecommunications Labeling, Electrical Labeling, General Identification, Laboratory Labeling
1 Tape/Cartridge
25 ft. x 0.5 in.
0.5 in.
0.5 in.