missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ FRIO-Temp™ Electronic Verification Lollipop Stem Thermometers for Ambient Rooms

For use in digital verification of sample temperatures in ambient calibration. H-B Instrument™ FRIO-Temp™ Electronic Verification Lollipop Stem Thermometers for Ambient Rooms render accurate temperature reading without sample contamination.

Brand:  H-B Instrument™ B60210-2200

 View more versions of this product

Product Code. 17560666

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Alert:

Not for use in ovens or freezers

This item is not returnable. View return policy

Description

Description

  • Entire digital unit can be placed directly in refrigerators, incubators and ambient rooms
  • Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish)
  • Individually serialized
    • Certifications

      ISO 17025, CE marked

Specifications

Specifications

Glass Beads
22°C Ambient
5.25 in.
3 in.
Stainless Steel
1.5 in.
70°C
0°C to 70°C
±0.5°C
60 mL Plastic Bottle
Digital verification of sample temperatures in ambient calibration
133 mm
75 mm
0.1°C
Digital Thermometer
0°C
Yes
38 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers