missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ Frontier™ 5000 Series Mini Centrifuge

Powerful and convenient mini-centrifuge for simple and quick life science separation applications.

Brand:  Ohaus™ 30134156

214.68 EUR valid until 2024-12-31
Use promo code "21929" to get your promotional price.


Product Code. 15806762

  • 274.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Small but powerful mini-centrifuge for simple and quick liquid separation.
  • Can accommodate 0.2 mL 8 strip tubes or 32 individual PCR tubes.
  • With the included adapter, 0.2 mL and 0.5 mL tubes can also be accommodated in the 8 place tube rotor.
  • Quiet, safe, and easy operation for your everyday quick spin-down applications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2,000 x g
Mini centrifuge with two rotors included, max RPM 6000, max RCF 2000 x g
Centrifuge, Mini, FC5306, EU
168.00 x 150.00 x 128.000 mm
168.00 mm
0 to 6,000 rpm
128.000 x 150.00 x 168.00 mm
Frontier
Angle 8 x 1.5 mL / 2.0 ml, Angle Rotor 4 x 8 Place PCR Strips
No Display
CEFCCTUV
100/240 V
6.60 in.
FC5306
Continuous
1.100 kg
120/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.