Learn More
CleanSpace Technology™ Full Face Mask Quantitative Fit Test PortaCount Adaptor
The CleanSpace Full Face Mask Quantitative Fit Test (PortaCount) Adaptor is designed to be used with the CleanSpace Full Face Mask (PAF-1014). It is utilised as a mask fit test adaptor to be fitted in the front of the mask for Quantitative Fit (TSI PortaCount) Testing.
Brand:  CleanSpace Technology™ PAF-1015
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15950082
  • 159.00 € / Each
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
Specifications
Obsolete
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
CleanSpace Technology™ Full Face Mask Quantitative Fit Test PortaCount Adaptor > For use with: CleanSpace Full Face Mask (PAF-1014)