missing translation for 'onlineSavingsMsg'
Learn More

CleanSpace™ Technology Full Face Mask Tear-Off Visor Anti-Scratch Protectors

Full face mask tear-off visor anti-scratch protectors are protective films for the full face mask visor.

Brand:  CleanSpace™ Technology PAF-1018

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15980082

  • 89.95 € / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pack of 10 tear offs
  • Clear and easy to use
  • Used with the revolutionary CleanSpace™ Respirator: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
Specifications

Specifications

AS/NZS1716: 2012, EN 12942
Polyester
PAF-1014, PAF-1106
10/Pk.
210 x 125 mm
Single-use
Tear off Visor Anti-Scratch Protectors
PAF-1018
Store at -10°C to +55°C
75 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.