Learn More
3M™ Full Facepiece Reusable Respirator
3M™ Promask™ Full Facepiece Reusable Respirator FM3 is a multi-function full-face mask designed to provide durable and comfortable protection against a wide range of respiratory hazards. Use in conjunction with approved 3M powered air systems or as a negative pressure respirator.
915.00 €
Specifications
Connection Type Bayonet & DIN
Color Black
Product Type Full Face Mask
Suspension Type 5 Point Strap
View More Specs
Products 2
Product Code Brand Size Model Price Quantity & Availability  
Product Code Brand Size Model Price Quantity & Availability  
17786973
View Documents
3M™
7100237033
Small FF-301
915.00 €
Pack of 4
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
17766973
View Documents
3M™
7100236989
Large FF-302
915.00 €
Pack of 4
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
Description
Description
  • Use as a negative-pressure single-filter mask or as a face piece for a powered air respirator
  • Sweat port helps to provide user comfort during long periods of use
  • Visor provides a good field of view and comes in choice of polycarbonate or high-heat material
  • Side-fitting filter port and clear inner mask for good forwards and downwards vision
  • Filter doesn't interfere with front-mounted speech diaphragm to aid in easy communication
Specifications
Specifications
Bayonet & DIN
Black
Full Face Mask
5 Point Strap
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
3M™ Full Facepiece Reusable Respirator