missing translation for 'onlineSavingsMsg'
Learn More

Fullerene powder, 99+% C{70}

Product Code. 11389218
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11389218

Brand: Thermo Scientific Alfa Aesar 042600.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115383-22-7 | C70 | 840.77 g/mol

Fullerene powder is used for biomedical applications including the design of high-performance magnetic resonance imaging (MRI) contrast agents, X-ray imaging contrast agents, photodynamic therapy and drug and gene delivery. It is used in superconductors, photoactive polymers, film material modifiers, abrasive agent for cutting and grinding applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115383-22-7
Molecular Formula C70
Molecular Weight (g/mol) 840.77
MDL Number MFCD00282904
InChI Key ATLMFJTZZPOKLC-UHFFFAOYSA-N
Synonym fullerene, fullerene 70, rugbyballene, carbon, 70-d5h fullerene, buckminsterfullerene, 5,6 fullerene-c70-d5h 6, c70-d5h 6 5,6 fullerene, fullerene powder, c 250mg
PubChem CID 16131935
ChEBI CHEBI:33195
IUPAC Name (C\{70}-D\{5h(6)})[5,6]fullerene
SMILES C12=C3C4=C5C6=C7C8=C9C%10=C%11C%12=C%13C%10=C%10C8=C5C1=C%10C1=C%13C5=C8C1=C2C1=C3C2=C3C%10=C%13C%14=C3C1=C8C1=C3C5=C%12C5=C8C%11=C%11C9=C7C7=C9C6=C4C2=C2C%10=C4C(=C29)C2=C6C(=C8C8=C9C6=C4C%13=C9C(=C%141)C3=C85)C%11=C27

Specifications

Color Gray to Black
Density 1.7 g/mL
Boiling Point 500°C to 600°C (sublimation)
Quantity 250 mg
UN Number UN1325
Solubility Information Soluble in benzene and toluene. Insoluble in water.
Percent Purity ≥99%
Physical Form Powder
Assay C70
Chemical Name or Material Fullerene powder
compliance-icons compliance-icons
Produktidentifikator
  • Fullerene powder C70
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplerende oplysninger
  • MIXTURE LIST-Contains : Fullerene C70

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.