missing translation for 'onlineSavingsMsg'
Learn More
Fullerene powder, hydroxylated, C{60}(OH)∣n,, Thermo Scientific Chemicals Product Code.: 11339557

Fullerene powder, hydroxylated, C{60}(OH)∣n,, Thermo Scientific Chemicals

Product Code. 11339557
100 mg
Click to view available options
Quantity:
25 mg
100 mg
This item is not returnable. View return policy

Product Code. 11339557

Brand: Thermo Scientific Alfa Aesar 041182.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99685-96-8
Molecular Formula C60
Molecular Weight (g/mol) 720.66
MDL Number MFCD06202029
InChI Key XMWRBQBLMFGWIX-UHFFFAOYSA-N
Synonym fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
PubChem CID 123591
ChEBI CHEBI:33128
IUPAC Name (C\{60}-I\{h})[5,6]fullerene
SMILES C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41

Specifications

Quantity 100 mg
UN Number UN1325
Physical Form Powder
Chemical Name or Material Fullerene powder, Hydroxylated, C60(OH)n

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD27P23 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041182.MB,041182.MC
Lot NumberI15T030 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041182.MB,041182.MC
Lot NumberH20M01 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041182.MB,041182.MC
Lot NumberB09R005 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number041182.MB,041182.MC
Lot NumberH29T010 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number041182.MB,041182.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fullerene powder, hydroxylated, C{60}(OH)∣n,, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.