missing translation for 'onlineSavingsMsg'
Learn More

Fulvestrant, MedChemExpress

MedChemExpress Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant is also a GPR30 agonist. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy.

79.65 € - 367.20 €

Specifications

Chemical Name or Material Fulvestrant
CAS 129453-61-8
Color White
Purity Grade Notes Research
Molecular Formula C32H47F5O3S
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16455788
Encompass_Preferred
View Documents
Medchem Express
HY-13636
10 mg
79.65 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16406266
Encompass_Preferred
View Documents
Medchem Express
HY-13636
100 mg
367.20 €
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16401947
Encompass_Preferred
View Documents
Medchem Express
HY-13636
50 mg
201.15 €
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fulvestrant (ICI 182780) is a pure antiestrogen and a potent estrogen receptor (ER) antagonist with an IC50 of 9.4 nM. Fulvestrant is also a GPR30 agonist. Fulvestrant effectively inhibits the growth of ER-positive MCF-7 cells with an IC50 of 0.29 nM. Fulvestrant also induces autophagy and has antitumor efficacy.
Specifications

Specifications

Fulvestrant
White
C32H47F5O3S
DMSO : 250 mg/mL (412.02 mM; Need ultrasonic)
606.77
95.0%
Solid
129453-61-8
Research
ICI 182780 ZD 9238 ZM 182780
C[C@@]12[C@@H](O)CC[C@@]1([H])[C@]3([H])[C@H](CCCCCCCCCS(CCCC(F)(F)C(F)(F)F)=O)CC4=C(C=CC(O)=C4)[C@@]3([H])CC2
606.77
Research
Cancer-programmed cell death
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only