missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Safety Funnel 'ARNOLD' with Ball and Lid, V2.0, HDPE-EC

The strong and electrically conductive safety funnel ARNOLD V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms.

211.00 € - 322.00 €

Specifications

Diameter (Metric)Top 200 mm (Funnel)
Material HDPE-EC
Color Black
For Use With (Application) Safe Disposal of Liquid and Flammable Wastes
Product Type Safety Funnel
View More Specs

Products 15
Product Code Brand Includes Stopper Number Product Type Price Quantity & Availability  
Product Code Brand Includes Stopper Number Product Type Price Quantity & Availability  
17142235
View Documents Promotion Details
SCAT™
317627
Lance (220 mm), Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve B83 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17192225
View Documents Promotion Details
SCAT™
317622
Lance (220 mm), Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve GL45 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17465053
View Documents Promotion Details
SCAT™
611542
Safety Funnel with Hinged Lid GL45 Safety Funnel This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17288860
View Documents Promotion Details
SCAT™
317633
Splash Guard, Removable Sieve, Grounding Cable, Lid GL45 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17102245
View Documents Promotion Details
SCAT™
317638
Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve GL45 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17172235
View Documents Promotion Details
SCAT™
317630
Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve R2 Safety Funnel
211.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17162235
View Documents Promotion Details
SCAT™
317629
Lance (220 mm), Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve S50 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17112235
View Documents Promotion Details
SCAT™
317624
Lance (220 mm), Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve S51 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17122235
View Documents Promotion Details
SCAT™
317625
Lance (220 mm), Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve S55 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17182225
View Documents Promotion Details
SCAT™
317621
Lance (220 mm), Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve S60/61 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17298860
View Documents Promotion Details
SCAT™
317634
Splash Guard, Removable Sieve, Grounding Cable, Lid S60/61 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17132235
View Documents Promotion Details
SCAT™
317626
Lance (220 mm), Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve S65 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17152235
View Documents Promotion Details
SCAT™
317628
Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve S70/71 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17102235
View Documents Promotion Details
SCAT™
317623
Lance (220 mm), Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve S90 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
17192235
View Documents Promotion Details
SCAT™
317635
Lance (220 mm), Splash Guard, Removable Sieve, Grounding Cable, Lid, Ball Valve S95 Safety Funnel
243.00 €
Each
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
Description

Description

The safety funnel ARNOLD V2.0 with lid and ball valve is available for different container threads. In order to be able to produce in an environmentally friendly and economical way, we supply individual variants with a thread adapter, which has no influence on the functionality or safety of the funnel.

  • Two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic or explosive gases
  • Electrically conductive HDPE material with grounding connection to prevent flying sparks
  • Hinged lid with sealing ring for closure when the funnel is not in use
  • Optimized sieve for collecting larger particles and for depositing laboratory bottles
  • Improved splash guard with integrated air channels to prevent bubble formation
  • Ball valve with automatic opening mechanism during filling
  • Individually shortenable safety lance for clean drainage of liquid waste
  • All major components are removable for easy cleaning
Specifications

Specifications

200 mm (Funnel)
Black
Safety Funnel
HDPE-EC
Safe Disposal of Liquid and Flammable Wastes
Videos
SDS
Documents

Documents

Product Certifications