Learn More
SCAT™ Funnel with Lid and Stainless Steel Strainer
Open containers create fire and health hazards. Collect liquid waste safely and according to the latest state of the art: This funnel is characterised by its large diameter of 250 mm.
Brand: SCAT™ 108670
This item is not returnable.
View return policy
Description
- The stainless steel strainer safely catches solids such as magnetic stirring bars.
- With male thread R2'' BSP/G2'' / BCS 56x4
Specifications
250 mm | |
Black | |
Funnel |
HDPE | |
Funnel made of PE-HD in red with sieve from stainless steel |