missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Long Stem Funnel

Achieve quick and efficient filtration for food-related contents. Kartell™ Long Stem Funnel is ideal for fast filtration using standard filter papers.

Brand:  Kartell™ 0016200

Product Code. 12675686

  • 82.05 € / Pack of 10
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The 60° angles sides are provided with ribs for fast filtration
  • Allow the use of standard size filter papers
  • Suitable for foodstuff
Specifications

Specifications

65mm
8mm
Natural
50 mL
Polypropylene
150mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.