missing translation for 'onlineSavingsMsg'
Learn More

Furegrelate Sodium Salt, ≥99%, Thermo Scientific Chemicals

Product Code. 15403989
Click to view available options
Quantity:
5 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibitor of thrombaxane synthetase

Inhibitor of thrombaxane synthetase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85666-17-7
Molecular Formula C15H10NNaO3
Synonym U-63557A; Sodium furegrelate

Specifications

Quantity 50 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Inhibitor of thrombaxane synthetase
Formula Weight 275.20g/mol
Solubility Soluble in water at approximately 15mg/ml
Product Type Signal Transduction Reagent
Form Crystalline solid
Assay ≥99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'