Learn More
Furfurylamine, 99+%, Thermo Scientific Chemicals
22.01 € - 215.07 €
Chemical Identifiers
CAS | 617-89-0 |
---|---|
Molecular Formula | C5H7NO |
Molecular Weight (g/mol) | 97.12 |
MDL Number | MFCD00003258 |
InChI Key | DDRPCXLAQZKBJP-UHFFFAOYSA-N |
Synonym | furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl |
PubChem CID | 3438 |
IUPAC Name | furan-2-ylmethanamine |
SMILES | C1=COC(=C1)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10244820
|
Thermo Scientific Acros
119800050 |
5 mL | Glass bottle |
22.01 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10616502
|
Thermo Scientific Acros
119802500 |
250 mL | Glass bottle |
60.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10307040
|
Thermo Scientific
119800010 |
1 L | Glass bottle |
215.07 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
617-89-0 | |
97.12 | |
DDRPCXLAQZKBJP-UHFFFAOYSA-N | |
3438 | |
C1=COC(=C1)CN |
C5H7NO | |
MFCD00003258 | |
furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl | |
furan-2-ylmethanamine |
Specifications
617-89-0 | |
Brown-Yellow to Colorless | |
145°C to 146°C (761.0 mmHg) | |
Authentic | |
C5H7NO | |
MFCD00003258 | |
1.09 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether | |
C1=COC(=C1)CN | |
97.12 | |
1.6 mPa.s (20°C) | |
99+% | |
Furfurylamine, 99+% |
-70°C | |
1.0900g/mL | |
37°C | |
99+% | |
1.4890 to 1.491 | |
18, 584 | |
furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl | |
DDRPCXLAQZKBJP-UHFFFAOYSA-N | |
furan-2-ylmethanamine | |
3438 | |
97.12 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-536-9
missing translation for 'rtecsNumber' : LU9275000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.