Learn More
Carbolite™ FuRNAce Bwf11/13
Brand:  Carbolite™ CWF-B 11/13
Product Code. 12973751
  • 6260.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ashing and burn off furnace requires controller 201PID or 2416CG or 2408CP (specify with order) 230V 50/60Hz 1100°C 12.9L Carbolite
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products