Learn More
Carbolite™ Furnace with Uniform Temperature Envelope
Brand:  Carbolite™ CWF1323-230SN+&02-301
 View more versions of this product
Product Code. 12953741
  • 10780.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
301 PID Controller
1300°C
400 x 245 x 235 mm
254 mm
235 mm
400 mm
230 V
50/60 Hz
23 L
Furnace
505 mm
705 mm
675 mm
230 V 50/60 Hz 7000 w
7000 w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products