missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fursultiamine, MedChemExpress
MedChemExpress Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.
40.50 € - 238.95 €
Specifications
Chemical Name or Material | Fursultiamine |
---|---|
CAS | 804-30-8 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C17H26N4O3S2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16498728
|
Medchem Express
HY-B2082 |
10 mg |
59.40 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16478836
|
Medchem Express
HY-B2082 |
100 mg |
238.95 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16433816
|
Medchem Express
HY-B2082 |
25 mg |
113.40 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16424647
|
Medchem Express
HY-B2082 |
50 mg |
166.05 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16421148
|
Medchem Express
HY-B2082 |
5 mg |
40.50 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fursultiamine is a vitamin B1 derivative, has anti-nociceptive and antineoplastic activity. Fursultiamine can be used for vitamin B1 deficiency, osteoarthritis (OA) and cancer research.Spécification
Fursultiamine | |
White | |
C17H26N4O3S2 | |
O=CN(CC1=CN=C(C)N=C1N)/C(C)=C(SSCC2OCCC2)\CCO | |
398.54 | |
Research | |
COVID-19-immunoregulation |
804-30-8 | |
Research | |
DMSO : 106.7 mg/mL (267.73 mM; Need ultrasonic and warming) | |
398.54 | |
99.86% | |
Solid |
Sécurité et traitement
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only