missing translation for 'onlineSavingsMsg'
Learn More

Fusion Mixture, Certified AR for Analysis, Fisher Chemical™

123.00 € - 556.00 €

Chemical Identifiers

MDL Number 146979
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10316140
View Documents Promotion Details
Fisher Chemical
F/2250/53
500 g
556.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
10616262
View Documents Promotion Details
Fisher Chemical
F/2250/60
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

146979

Specifications

White
146979
Plastic powder jar
Solid
122.1g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Fusion mixture

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications