Learn More
Asecos™ FX-CLASSIC-90 Recirculating Air Safety Storage Cabinet
For the safe and approved storage of flammable liquids and further hazardous substances with air circulation for the extraction and filtering of vapors that are harmful to health.
Brand:  Asecos™ 3328604133656
Product Code. 15630159
  • 7665.00 € / Each
Explore more special offers
Description
Description
  • Safety for employees due to avoidance of explosive atmospheres
  • Fire resistance of 90 minutes
  • Maximum flexibility in installation, storage and available interior equipment
  • Recirculating air filter cabinets enable the common storage, extraction and filtration of the most diverse hazardous materials in one cabinet
  • Electronic monitoring for filter saturation, air recirculation rate, service intervals and manual filter check
  • Eco-friendly with long filter lifetimes and reusable filter system
  • Possible interior equipment includes drawers, height-adjustable shelves and bottom collecting sumps
  • Robust construction and longevity – triple-hinged doors, safety elements assembled outside of storage compartments for increased corrosion protection, scratch- and impact-resistant surface and easy to clean
  • Doors stop in any position and are self-closing in the event of fire
  • No unauthorized use – doors lockable with cylinder locking (integration into an existing locking system possible) with locking state indicator (red/green)
  • In accordance with EN 14470-1
  • Easy transport with integrated base with removable, optional base cover
  • Easy alignment with adjusting aids to compensate for uneven floors
  • Ideal for the direct installation in working areas with a very low noise level, approx. 39 dB
  • With multi-stage wideband filter
Specifications
Specifications
EN 14470-1
522 x 450 x 1647 mm
Fire-rated (1.5 hr.)
Air circulation, filtering, extraction
2
Light Gray
3 x powder-coated sheet steel shelves, load capacity 75 kg; 1 x powder-coated sheet steel perforated insert, load capacity 75 kg; 1 x powder-coated sheet steel bottom collecting sump, capacity 22 L; 3 x PP tray shelves, load capacity 50 kg, capacity 11 L; 1 x PP bottom collecting sump, capacity 11 L
615 x 1193 x 2294 mm
Storage of flammable liquids and hazardous substances
600 kg
565.5 kg
Safety storage cabinet