missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ FxCycle™ Far Red Stain

FxCycle™ Far Red Stain

Brand:  Molecular Probes™ F10348

Product Code. 10003774

  • 534.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

FxCycle™ Far Red stain (F10348) is used for flow cytometric analysis of DNA content in fixed cells when combined with RNAse. This dye takes advantage of the commonly available 633⁄5 nm excitation sources with emission around 660 nm. With multicolor cell cycle studies possible using flow cytometry, it is often necessary to analyze DNA content on alternative lasers to preserve the common 488 nm laser and detection channels for other markers. Well suited for the popular red laser, FxCycle™ Far Red stain is a good choice for DNA content analysis in multicolor cell cycle studies.

View a selection guide for all products related to cell cycle analysis of fixed and live cells in flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 5 vials of FxCycle™ far red stain. Store at ≤-20°C and protected from light.
Flow Cytometer
FxCycle™ Far Red Stain
Liquid
1 kit
DMSO (Dimethylsulfoxide)
Fluorescence
Stain
633
FxCycle™, Molecular Probes™
Room Temperature
Nucleic Acids
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.