Learn More
Asecos™ G-OD Outdoor Safety Storage Cabinet
For the safe and approved outdoor storage of flammable or toxic gases.
Brand:  Asecos™ 30692015
 View more versions of this product
Product Code. 15516895
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Robust, high-quality construction perfect for installation in outdoor areas
  • Completely galvanized sheet steel construction with plastic laminated structured surface and stainless steel base
  • Sloping roof with supernatant allows water to drain
  • Wing doors with a large opening angle for ease of access
  • Solid 3-point locking with lockable turning lever (also available as profile cylinder lock)
  • Effective ventilation through openings beneath the doors and at the rear wall
  • Comfortable adjusting possibilities – integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors
  • In accordance with TRGS 510
Specifications
Specifications
TRGS 510
370 x 1345 x 2070 mm
Wing
No Window
Plastic laminated
Light Gray
Safety storage cabinet
Tall cabinet with wing doors
400 x 1356 x 2149 mm
Outdoor storage of gas cylinders
600 kg
161 kg
Steel (Galvanized)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Asecos™ G-OD Outdoor Safety Storage Cabinet > Tall cabinet with wing doors