Learn More
G3BP1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E4A2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66486-1-IG-150UL
 View more versions of this product
Product Code. 16858405
  • 410.00 € / 150µL
Explore more special offers
Description
Description
May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves c-myc mRNA preferentially at the 3′UTR. ATP- and magnesium-dependen
Specifications
Specifications
G3BP1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P97855, Q13283
G3bp1
G3BP1 Fusion Protein ag3728
150 μL
Primary
Human, Mouse, Pig, Rat
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1E4A2
Unconjugated
G3BP1
G3BP, G3BP 1, G3BP1, HDH VIII
Mouse
Protein G
RUO
100513766, 10146, 171092, 27041
-20°C