missing translation for 'onlineSavingsMsg'
Learn More

Corning™ G418 Sulfate, Powder
BIOPHARMA

Packaged in plastic bottles

432.06 € - 4473.18 €

Specifications

Content And Storage 2° to 8°C
Form Powder
Product Type Antibiotic
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15313771
BIOPHARMA
View Documents
Corning™
61-234-RG
5 g
432.06 €
5g
Please to purchase this item. Need a web account? Register with us today!
15323771
BIOPHARMA
View Documents
Corning™
61-234-RK
50 g
4473.18 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

2° to 8°C
Powder
Antibiotic
Videos
SDS
Documents

Documents

Product Certifications