missing translation for 'onlineSavingsMsg'
Learn More

G418 Sulfate Solution, MP Biomedicals

50mg/ml active antibiotic sterile-aqueous solution. Purity: >>98%

236.00 € - 402.00 €

Specifications

Type G418 Sulfate
Chemical Name or Material G418 Sulfate Solution

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11430435
View Documents
MP Biomedicals
091672546
20 mL
236.00 €
20mL
Please to purchase this item. Need a web account? Register with us today!
11440435
View Documents
MP Biomedicals
091672548
50 mL
402.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

G418 Sulfate
G418 Sulfate Solution
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.