missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAPL1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E10F6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66458-1-IG-20UL
This item is not returnable.
View return policy
Description
Increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.Specifications
GABARAPL1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q0VGK0, Q8R3R8, Q9H0R8 | |
Gabarapl1 | |
GABARAPL1 Fusion Protein Ag1473 | |
20 μL | |
Primary | |
Mouse, Rat, Porcine, Human | |
Antibody | |
IgG1 |
Western Blot | |
1E10F6 | |
Unconjugated | |
GABARAPL1 | |
APG8 LIKE, APG8L, ATG8, ATG8L, GABARAPL1, GEC 1, GEC1 | |
Mouse | |
Protein G | |
RUO | |
100462751, 23710, 57436, 689161 | |
-20°C | |
Liquid |