missing translation for 'onlineSavingsMsg'
Learn More

Gabexate mesylate, Tocris Bioscience™

Product Code. 18083932
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Serine protease inhibitor; inhibits thrombin, trypsin, kallikrein and plasmin

Gabexate mesylate is a serine protease inhibitor; Gabexate mesylate inhibits trypsin, plasmin, plasma kallikrein and thrombin (IC50 values are 9.4, 30, 41 and 110 μM respectively). Antithrombotic in vitro and in vivo. Gabexate mesylate also inhibits LPS-induced TNF-α production, probably by inhibiting NF-κB and AP-1 activation. Gabexate mesylate attenuates allergen-induced airway inflammation and eosinophils infiltration in the lung of asthma murine model.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56974-61-9
Molecular Formula C17H27N3O7S
Molecular Weight (g/mol) 417.48
MDL Number MFCD00210299
InChI Key DNTNDFLIKUKKOC-UHFFFAOYSA-N
Synonym gabexate mesylate, gabexate mesilate, gabexate monomethanesulfonate, megacart, gabexate methanesulfonate, foy, dsstox_cid_25591, dsstox_rid_80986, dsstox_gsid_45591
PubChem CID 6604561
IUPAC Name ethyl 4-[(6-{[amino(azaniumyl)methylidene]amino}hexanoyl)oxy]benzoate methanesulfonate
SMILES CS([O-])(=O)=O.CCOC(=O)C1=CC=C(OC(=O)CCCCCN=C(N)[NH3+])C=C1

Specifications

Quantity 10 mg
Formula Weight 417.48
Percent Purity >98%
Chemical Name or Material Gabexate mesylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.