missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Antibody (9E10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 33 publications
197.00 € - 548.00 €
Specifications
| Classification | Monoclonal |
|---|---|
| Form | Purified |
| Isotype | IgG1 κ |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18740444
|
Novus Biologicals
NB600-302SS |
197.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18717933
|
Novus Biologicals
NB600-302 |
0.1 mL |
548.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-Myc Monoclonal specifically detects c-Myc in Human, Mouse, Bovine, Drosophila samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, Sandwich ELISA, Flow (Intracellular).Specifications
| Monoclonal | |
| Purified | |
| IgG1 κ | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title