missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) bromide, ultra dry, 99.99% (metals basis)

Product Code. 11395897
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11395897

Brand: Thermo Scientific Alfa Aesar 043673.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13818-75-2 | GdBr3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13818-75-2
Molecular Formula GdBr3
MDL Number MFCD00049741

Specifications

Melting Point 770°C
Physical Form Powder
Quantity 100 g
Solubility Information It is soluble in water. Also soluble in THF,1,2-Diethoxyethane,p-dioxane,and alkyl amine.
Formula Weight 396.96
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Gadolinium (III) bromide, Ultra dry

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.