missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals

Product Code. 10412683
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10138-52-0
Molecular Formula Cl3Gd
Molecular Weight (g/mol) 263.61
MDL Number MFCD00011024
InChI Key MEANOSLIBWSCIT-UHFFFAOYSA-K
Synonym gadolinium chloride, gadolinium trichloride, gadolinium chloride gdcl3, gdcl3, gadolinium iii chloride, gadolinium 3+ chloride, wln: gd g3, gadolinium iii chloride, ultra dry, gadolinium iii chloride, anhydrous, ?gdcl3
PubChem CID 61486
ChEBI CHEBI:37288
IUPAC Name trichlorogadolinium
SMILES Cl[Gd](Cl)Cl

Specifications

Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 58.2 to 61.1% (Gd) (Complexometry)
Linear Formula GdCl3
Solubility Information Solubility in water: soluble
Formula Weight 263.61
Percent Purity 99.90%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Gadolinium(III) chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.