missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Gadolinium(III) chloride hexahydrate is employed in synthesis and NMR studies of the paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes with possible use as contrast agents for magnetic resonance imaging..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Physical Form | Crystalline Aggregates |
Quantity | 25 g |
Merck Index | 14,4326 |
Solubility Information | soluble in water. |
IUPAC Name | gadolinium(3+) hexahydrate trichloride |
Formula Weight | 371.70 (263.61 Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Odor | Characteristic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'