missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11304118
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For vacuum deposition

Gadolinium(III) fluoride, anhydrous is employed in the synthesis of non-oxide glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13765-26-9
Molecular Formula F3Gd
Molecular Weight (g/mol) 214.25
MDL Number MFCD00016113
InChI Key TYIZUJNEZNBXRS-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) trifluoride
SMILES [F-].[F-].[F-].[Gd+3]

Specifications

Melting Point 1231°C
Color White
Physical Form Pieces
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 214.25
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.1 g/mL
Assay (REO)
Chemical Name or Material Gadolinium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'