missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) hydroxide hydrate, 99.9% (REO), Thermo Scientific Chemicals

73.97 € - 285.42 €

Chemical Identifiers

CAS 100634-91-1
Molecular Formula GdH3O3
Molecular Weight (g/mol) 208.27
MDL Number MFCD00049930
InChI Key ILCLBMDYDXDUJO-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Gd+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398488
SDS View Documents
Thermo Scientific Alfa Aesar
040507.06
5 g
73.97 €
5g
Please to purchase this item. Need a web account? Register with us today!
11388488
SDS View Documents
Thermo Scientific Alfa Aesar
040507.14
25 g
285.42 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100634-91-1
208.27
ILCLBMDYDXDUJO-UHFFFAOYSA-K
[OH-].[OH-].[OH-].[Gd+3]
GdH3O3
MFCD00049930
gadolinium(3+) trihydroxide

Specifications

100634-91-1
GdH3O3
14,4326
[OH-].[OH-].[OH-].[Gd+3]
208.27
99.9%
Gadolinium(III) hydroxide hydrate
Powder
MFCD00049930
ILCLBMDYDXDUJO-UHFFFAOYSA-K
gadolinium(3+) trihydroxide
208.27 (Anhydrous)
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 240-516-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) hydroxide hydrate, 99.9% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.