missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

138.69 € - 488.43 €

Chemical Identifiers

CAS 13572-98-0
Molecular Formula GdI3
Molecular Weight (g/mol) 537.96
MDL Number MFCD00016114
InChI Key IZZTUGMCLUGNPM-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) triiodide
SMILES [I-].[I-].[I-].[Gd+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338607
SDS View Documents
Thermo Scientific Alfa Aesar
035735.03
1 g
138.69 €
1g
Please to purchase this item. Need a web account? Register with us today!
11328607
SDS View Documents
Thermo Scientific Alfa Aesar
035735.06
5 g
488.43 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a compound in semiconductors and other high purity applications, for contrast imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

13572-98-0
537.96
IZZTUGMCLUGNPM-UHFFFAOYSA-K
[I-].[I-].[I-].[Gd+3]
GdI3
MFCD00016114
gadolinium(3+) triiodide

Specifications

926°C
Powder
MFCD00016114
IZZTUGMCLUGNPM-UHFFFAOYSA-K
gadolinium(3+) triiodide
537.96
Ampule
(REO)
13572-98-0
GdI3
Soluble in water.
[I-].[I-].[I-].[Gd+3]
537.96
99.99%
Hygroscopic
Gadolinium(III) iodide, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 236-997-6

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.