missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadolinium(III) nitrate hydrate, 99.5% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
It is applied as an oxidizing agent and the precursor material in the preparation of gadolinium compounds. It is extensively employed in the production of specialized glasses and ceramics. It is also employed in heavy water nuclear reactors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 92°C |
Color | White |
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN1477 |
Merck Index | 14,4326 |
Solubility Information | Soluble in water. |
Formula Weight | 343.26 (Anhydrous) |
Percent Purity | 99.9% |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'