missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

78.70 € - 791.00 €

Chemical Identifiers

CAS 94219-55-3
Molecular Formula GdN3O9
Molecular Weight (g/mol) 343.26
MDL Number MFCD00011023
InChI Key MWFSXYMZCVAQCC-UHFFFAOYSA-N
IUPAC Name gadolinium(3+) trinitrate
SMILES [Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11321009
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011284.06
5 g
78.70 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11341009
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011284.14
25 g
217.00 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11331009
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011284.22
100 g
791.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gadolinium(III) nitrate is used as an oxidizing agent and the precursor material in the preparation of gadolinium compounds. It is extensively employed in the production of specialized glasses and ceramics. It is also employed in heavy water nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

94219-55-3
343.26
MWFSXYMZCVAQCC-UHFFFAOYSA-N
[Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
GdN3O9
MFCD00011023
gadolinium(3+) trinitrate

Specifications

92°C
White
GdN3O9
UN1477
Soluble in water.
[Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
343.26
99.99%
Hygroscopic
(REO)
94219-55-3
Crystalline Aggregates
MFCD00011023
14,4326
MWFSXYMZCVAQCC-UHFFFAOYSA-N
gadolinium(3+) trinitrate
343.26 (Anhydrous)
Reacton™
2.406 g/mL
Gadolinium(III) nitrate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

missing translation for 'rtecsNumber' : LW4550000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only