Learn More
Gadolinium(III) oxalate hydrate, 99.9% (REO), Thermo Scientific™
35.70 € - 103.00 €
Chemical Identifiers
CAS | 100655-00-3 |
---|---|
Molecular Formula | C6H8Gd2O13 |
Molecular Weight (g/mol) | 602.62 |
MDL Number | MFCD00150145 |
InChI Key | FEIDMMDEDUAFJQ-UHFFFAOYSA-N |
Synonym | oxalic acid-gadolinium-water 3/2/1 |
PubChem CID | 92020445 |
IUPAC Name | gadolinium;oxalic acid;hydrate |
SMILES | O.[Gd].[Gd].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397118
|
Thermo Scientific Alfa Aesar
015308.14 |
25 g |
35.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387118
|
Thermo Scientific Alfa Aesar
015308.22 |
100 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
100655-00-3 | |
602.62 | |
FEIDMMDEDUAFJQ-UHFFFAOYSA-N | |
92020445 | |
O.[Gd].[Gd].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O |
C6H8Gd2O13 | |
MFCD00150145 | |
oxalic acid-gadolinium-water 3/2/1 | |
gadolinium;oxalic acid;hydrate |
Specifications
(decomposition) | |
100655-00-3 | |
Powder | |
Gd2(C2O4)3·xH2O | |
UN3288 | |
FEIDMMDEDUAFJQ-UHFFFAOYSA-N | |
gadolinium;oxalic acid;hydrate | |
92020445 | |
99.9% | |
(REO) |
110°C (in -6H2O) | |
White | |
C6H8Gd2O13 | |
MFCD00150145 | |
oxalic acid-gadolinium-water 3/2/1 | |
O.[Gd].[Gd].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O | |
602.62 | |
578.56 (Anhydrous) | |
Hygroscopic | |
Gadolinium(III) oxalate hydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures