missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) oxide, nanopowder, 99.99+% (REO), Thermo Scientific Chemicals

161.00 € - 1926.00 €

Chemical Identifiers

CAS 12064-62-9
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.50
MDL Number MFCD00011026
InChI Key CMIHHWBVHJVIGI-UHFFFAOYSA-N
Synonym gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
PubChem CID 159427
IUPAC Name digadolinium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Gd+3].[Gd+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11400650
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044020.14
25 g
161.00 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11325098
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044020.22
100 g
509.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11335098
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044020.36
500 g
1926.00 €
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gadolinium(III) oxide is used as a raw material for various fluorescent compounds, absorption materials in atomic reactions, nuclear fuels, magnetic bubble material and screen-sensitivity increasing material. It is also used in glass and electronic industries. It also acts as a neutron shield, a catalyst, a dielectric ceramic and is used for filament coatings, special glasses, lasers, masers and telecommunication. It also serves as a laboratory reagent. It plays an important role as a potential contrast agent for magnetic resonance imaging (MRI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12064-62-9
362.50
CMIHHWBVHJVIGI-UHFFFAOYSA-N
159427
[O--].[O--].[O--].[Gd+3].[Gd+3]
Gd2O3
MFCD00011026
gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
digadolinium(3+) trioxidandiide

Specifications

2330°C
Powder
MFCD00011026
gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
CMIHHWBVHJVIGI-UHFFFAOYSA-N
digadolinium(3+) trioxidandiide
159427
≥99.99%
(REO)
12064-62-9
Gd2O3
14,4326
Soluble in acid. Insoluble in water.
[O--].[O--].[O--].[Gd+3].[Gd+3]
362.50
362.5
7.407 g/mL
Gadolinium (III) oxide
Videos
Safety and Handling

Safety and Handling

P264b-P280i-P305+P351+P338

H319

missing translation for 'einecsNumber' : 235-060-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only